Merck
  • Determination of campesterol, stigmasterol, and beta-sitosterol in saw palmetto raw materials and dietary supplements by gas chromatography: collaborative study.
Determination of campesterol, stigmasterol, and beta-sitosterol in saw palmetto raw materials and dietary supplements by gas chromatography: collaborative study.
Journal of AOAC International (2007-06-22)
Wendy R Sorenson, Darryl Sullivan
ABSTRACT
An interlaboratory study was conducted to evaluate a method for the determination of campesterol, stigmasterol, and beta-sitosterol in saw palmetto raw materials and dietary supplements at levels >1.00 mg/100 g based on a 2-3 g sample. Test samples were saponified at high temperature with ethanolic KOH solution. The unsaponifiable fraction containing phytosterols (campesterol, stigmasterol, and beta-sitosterol) was extracted with toluene. Phytosterols were derivatized to trimethylsilyl ethers and then quantified by gas chromatography with hydrogen flame ionization detection. Twelve blind duplicates, one of which was fortified, were successfully analyzed by 10 collaborators. Recoveries were obtained for the sample that was fortified. The results were 99.8, 111, and 111% for campesterol, stigmasterol, and beta-sitosterol, respectively. For repeatability, the relative standard deviation (RSDr) ranged from 3.93 to 17.3% for campesterol, 3.56 to 22.7% for stigmasterol, and 3.70 to 43.9% for beta-sitosterol. For reproducibility, the RSDR ranged from 7.97 to 22.6%, 0 to 26.7%, and 5.27 to 43.9% for campesterol, stigmasterol, and beta-sitosterol, respectively. Overall, the Study Director approved 5 materials with acceptable HorRat values for campesterol, stigmasterol, and beta-sitosterol ranging from 1.02 to 2.16.
Sigma-Aldrich
Transcreener® ADP2 TR-FRET Red Assay
Pricing and availability is not currently available.