Merck
  • Generation of two iPSC lines (KAUSTi001-A, KAUSTi002-A) from a rare high-grade Klinefelter Syndrome patient (49-XXXXY) carrying a balanced translocation t(4,11) (q35,q23).
Generation of two iPSC lines (KAUSTi001-A, KAUSTi002-A) from a rare high-grade Klinefelter Syndrome patient (49-XXXXY) carrying a balanced translocation t(4,11) (q35,q23).
Stem cell research (2020-12-01)
Maryam Alowaysi, Elisabetta Fiacco, Veronica Astro, Antonio Adamo
ABSTRACT
Klinefelter Syndrome (KS) is the most common aneuploidy in humans (prevalence: 85-250 per 100,000 born males) and is characterized by one or more supernumerary X-chromosomes (47-XXY, 48-XXXY and 49-XXXXY karyotypes). KS is a multisystemic disorder associated to multiple phenotypic features including cardiac abnormalities, infertility, mental retardation, diabetes and increased cancer risk. Using a non-integrative mRNAs reprogramming approach, we generated two iPSC lines 48-XXXY and 49-XXXXY from a non-mosaic 49-XXXXY KS patient carrying a balanced translocation t(4,11) (q35,q23). These iPSC lines provide a unique cellular platform to study the molecular mechanisms underlying KS pathophysiology.
Sigma-Aldrich
Monoclonal Anti-SOX2 antibody produced in mouse, clone 10F10, ascites fluid