Merck
  • GLP-1 secretion in acute ischemic stroke: association with functional outcome and comparison with healthy individuals.
GLP-1 secretion in acute ischemic stroke: association with functional outcome and comparison with healthy individuals.
Cardiovascular diabetology (2019-07-17)
Martin Larsson, Cesare Patrone, Mia von Euler, Jens J Holst, David Nathanson
ABSTRACT
Glucagon-like peptide-1 (GLP-1) treatment has been shown to reduce stroke incidence in diabetes and also to be neuroprotective in experimental stroke models. The prognostic value of endogenous levels of GLP-1 in the recovery phase after stroke remains to be elucidated. The aim of the study was to investigate the potential association between GLP-1 levels and functional outcome after stroke and to determine whether GLP-1 is altered in the acute phase of stroke compared to 3 months post stroke and to healthy controls. Fasting GLP-1 was measured on hospital day 2-4 in patients without previously known diabetes (n = 59) that received recombinant tissue plasminogen activator (rtPA) for ischemic stroke. Fasting GLP-1 was measured again after 3 months and neurologic outcome was measured as modified Rankin Scale (mRS). mRS ≥ 2 was considered as unfavorable outcome. A control group of healthy individuals (n = 27) was recruited and their fasting GLP-1 was measured. Fasting GLP-1 was higher in the patients that suffered a stroke compared to healthy controls (25.1 vs. 18.0 pmol/L; p = 0.004). The GLP-1 levels did not change significantly at the 3-month follow up OGTT (25.8 vs. 25.6; p = 0.80). There was no significant association between GLP-1 levels and unfavorable mRS (OR 1.03, 95% CI 0.95-1.12, p = 0.50). Endogenous GLP-1 levels in patients that recently suffered an ischemic stroke are higher than in healthy controls and remained unchanged at the 3 months follow-up, possibly indicating an elevation of the levels of GLP-1 already pre-stroke. However, no association between endogenous GLP-1 and functional outcome of stroke 3 months post stroke was found.
Sigma-Aldrich
Human GCG / Glucagon ELISA Kit
Pricing and availability is not currently available.