Merck
  • Changes in expression of calbindin 28 kDa in the small intestine of red kidney bean (Phaseolus vulgaris) lectin-treated suckling piglets.
Changes in expression of calbindin 28 kDa in the small intestine of red kidney bean (Phaseolus vulgaris) lectin-treated suckling piglets.
Polish journal of veterinary sciences (2013-08-27)
A Zacharko-Siembida, J L Valverde Piedra, M B Arciszewski
ABSTRACT
Calbindin (CB) is a calcium binding protein playing a role in calcium uptake and anti-apoptotic cellular protection. The expression of CB was immunohistochemically studied in the small intestine of normal and red bean kidney lectin-treated suckling piglets. In the duodenum and jejunum (but not ileum) of lectin-treated animals overexpression of CB was noted in chromogranin A-immunoreactive (CgA-IR) neuroendocrine (NE) cells. In both control and experimental group a small population of CB-IR NE cells exhibited the presence of somatostatin (but not serotonin, histamine or CRF). After the lectin treatment, an increased (however not statistically significant) immunoreactivity to CB was found in a small subpopulation of neurons of outer submucous (but not inner submucous and myenteric) plexus. It is suggested that there is a functional interaction between lectin administration and CB-expression in the porcine small intestine. Future studies will be needed to clarify this processes.