Merck
  • SHP-1 suppresses endotoxin-induced uveitis by inhibiting the TAK1/JNK pathway.
SHP-1 suppresses endotoxin-induced uveitis by inhibiting the TAK1/JNK pathway.
Journal of cellular and molecular medicine (2020-11-19)
Xiaonan Zhuang, Jun Ma, Sisi Xu, Zhongcui Sun, Rong Zhang, Meng Zhang, Gezhi Xu
ABSTRACT
We investigated how Src-homology 2-domain phosphatase-1 (SHP-1) regulates the inflammatory response in endotoxin-induced uveitis (EIU), and the signalling pathways involved. One week after intravitreal injection of short hairpin RNA targeting SHP-1 or SHP-1 overexpression lentivirus in rats, we induced ocular inflammation with an intravitreal injection of lipopolysaccharide (LPS). We then assessed the extent of inflammation and performed full-field electroretinography. The concentrations and retinal expression of various inflammatory mediators were examined with enzyme-linked immunosorbent assays and Western blotting, respectively. SHP-1 overexpression and knockdown were induced in Müller cells to study the role of SHP-1 in the LPS-induced inflammatory response in vitro. Retinal SHP-1 expression was up-regulated by LPS. SHP-1 knockdown exacerbated LPS-induced retinal dysfunction and increased the levels of proinflammatory mediators in the retina, which was abrogated by a c-Jun N-terminal kinase (JNK) inhibitor (SP600125). SHP-1 overexpression had the opposite effects. In Müller cells, the LPS-induced inflammatory response was enhanced by SHP-1 knockdown and suppressed by SHP-1 overexpression. SHP-1 negatively regulated the activation of the transforming growth factor-β-activated kinase-1 (TAK1)/JNK pathway, but not the nuclear factor-κB pathway. These results indicate that SHP-1 represses EIU, at least in part, by inhibiting the TAK1/JNK pathway and suggest that SHP-1 is a potential therapeutic target for uveitis.