Merck
  • Individual human serum differs in the amount of antibodies with affinity for pig fetal ventral mesencephalic cells and the ability to lyse these cells by complement activation.
Individual human serum differs in the amount of antibodies with affinity for pig fetal ventral mesencephalic cells and the ability to lyse these cells by complement activation.
Cell transplantation (2005-01-15)
Jan Koopmans, Aalzen de Haan, Elinda Bruin, Ieneke van der Gun, Henk van Dijk, Jan Rozing, Lou de Leij, Michiel Staal
ABSTRACT
Xenografting pig fetal ventral mesencephalic (pfVM) cells to repair the dopamine deficit in patients with Parkinson's disease is the focus of both experimental and clinical investigations. Although there have been marked advances in the experimental and even clinical application of these xenogeneic transplantations, questions regarding the host's xenospecific immune response remain unanswered. It has been shown that human serum is able to lyse pfVM tissue by both anti-gal-gal and non-anti-gal-gal antibodies by complement activation. The aim of this study was to investigate whether interindividual differences exist in the levels of pfVM cell-specific IgM and IgG subclass antibodies, their ability to lyse pfVM cells in vitro and the relationship between both. Pig fetal VM cells were incubated with heat-inactivated serum from 10 different individuals and binding of IgM antibodies and IgG subclass antibodies to pfVM cells was analyzed by flow cytometry. The ability to lyse pfVM cells was analyzed exposing 51Cr-labeled pfVM cells to fresh serum or isolated IgM and IgG from the same individuals and subsequent determination of released 51Cr from lysed cells. Strong differences were found between individuals in the levels of pfVM cell-specific IgM antibodies: antibody levels differed up to 40-fold. pfVM-specific IgG1 and IgG2 levels were only detectable in a few individuals. The ability to lyse pfVM cells ranged from negligible lysis up to 66.5% specific lysis. There was a strong correlation between the levels of individual pfVM-specific IgM antibodies and the ability to lyse pfVM cells in vitro. Isolated IgM, but not IgG, was able to lyse pfVM cells in the presence of complement. In conclusion, the interindividual differences in the levels of IgM with affinity for pfVM cells and their ability to lyse pfVM cells in vitro are considerable. Only few individuals possessed IgG1 and IgG2 subclass antibodies with affinity for pfVM. These findings may influence patient selection for porcine transplants and chances of graft survival in individual patients.