Merck
  • Study for the Diagnostic Screening of Paroxysmal Nocturnal Hemoglobinuria in Older Patients with Unexplained Anemia and/or Cytopenia.
Study for the Diagnostic Screening of Paroxysmal Nocturnal Hemoglobinuria in Older Patients with Unexplained Anemia and/or Cytopenia.
Clinical laboratory (2020-09-10)
Zehra N Ozdemir, Osman Ilhan, Gulsum Ozet, Mesude Falay, Mustafa Yenerel, Tulin Tuglular, Mehmet Turgut, Birol Guvenc, Ali Unal, Orhan Ayyildiz, Neslihan Andic, Abdullah Hacihanefioglu, Fahri Sahin, Mehmet Sencan, Ridvan Ali, Guner H Ozsan, Rahsan Yildirim, Eyup N Tiftik, Anil Tombak, Ozan Salim, Emin Kaya, Olga M Akay, Vahap Okan, Mustafa Pehlivan, Guray Saydam
ABSTRACT
Paroxysmal nocturnal hemoglobinuria (PNH) is a rare acquired hematopoietic stem cell disease that may lead to weakness and death of patients, if unrecognized and untreated. Although consensus guidelines were reviewed recently for the diagnostic screening of PNH with multi-parameter flow cytometry (FCM), until now, no study has investigated the efficiency of such clinical indications in older patients. Overall, 20 centers participated in the study and a total of 1,689 patients were included, 313 of whom were at geriatric age and 1,376 were aged 18 - 64 years. We evaluated the efficiency of consensus clinical indications for PNH testing using FCM in peripheral blood samples and compared the results of older patients and patients aged 18 - 64 years. PNH clones were detected positive in 7/313 (2.2%) of the older patients. Five (74.4%) of the patients with PNH clones had aplastic anemia, 1 had unexplained cytopenia, and 1 patient had myelodysplastic syndrome (MDS) with refractory anemia. PNH clones were not detected in any older patients who were screened for unexplained thrombosis, Coombs (-) hemolytic anemia, hemoglobinuria, and others (e.g., elevated lactate dehydrogenase (LDH), splenomegaly). We detected PNH clones in 55/1376 (4%) samples of the patients aged under 65 years. Forty-two (76.4%) patients with PNH clones had aplastic anemia, 2 patients had Coombs (-) hemolytic anemia, 3 patients had unexplained cytopenia, 1 patient had MDS with refractory anemia, 1 patient had hemoglobinuria, and 6 (10.9%) had others (e.g., elevated LDH, splenomegaly). PNH clones were not detected in any patients who were screened for unexplained thrombosis. There was no statistical difference between the geriatric population and patients aged 18 - 64 years in terms of clinical indications for PNH screening with FCM (p = 0.49). Our results showed that the current clinical indications for PNH screening with FCM were also efficient in older patients. We suggest that older patients with unexplained anemia, myelodysplastic syndrome with refractory anemia, and unexplained cytopenia should be screened for PNH with FCM to identify patients who would benefit from treatment.
Sigma-Aldrich
Biphenyl-4,4′-dithiol, 95%