Merck
  • Hypothalamic-pituitary-adrenal axis activation and immune regulation in heat-stressed sheep after supplementation with polyunsaturated fatty acids.
Hypothalamic-pituitary-adrenal axis activation and immune regulation in heat-stressed sheep after supplementation with polyunsaturated fatty acids.
Journal of dairy science (2014-05-06)
M Caroprese, M G Ciliberti, G Annicchiarico, M Albenzio, A Muscio, A Sevi
ABSTRACT
The aim of this study was to assess the effects of supplementation with polyunsaturated fatty acids from different sources on immune regulation and hypothalamic-pituitary-adrenal (HPA) axis activation in heat-stressed sheep. The experiment was carried out during the summer 2012. Thirty-two Comisana ewes were divided into 4 groups (8 sheep/group): (1) supplemented with whole flaxseed (FS); (2) supplemented with Ascophyllum nodosum (AG); (3) supplemented with a combination of flaxseed and A. nodosum (FS+AG); and (4) control (C; no supplementation). On d 22 of the experiment, cortisol concentrations in sheep blood were measured after an injection of ACTH. Cellular immune response was evaluated by intradermic injection of phytohemagglutinin (PHA) at 0, 15, and 30 d of the trial. Humoral response to ovalbumin (OVA) was measured at 0, 15, and 30 d. At 0, 15, and 30 d of the experiment, blood samples were collected from each ewe to determine production of T-helper (Th)1 cytokines (IL-12 and IFN-γ), and Th2 cytokines (IL-10, IL-4, IL-13), and concentrations of heat shock proteins (HSP) 70 and 90. Ewes supplemented with flaxseed alone had greater cortisol concentrations and a longer-lasting cell-mediated immune response compared with ewes in the control and other groups. Anti-OVA IgG concentrations increased in all groups throughout the trial, even though ewes in the FS+AG group had the lowest anti-OVA IgG concentrations at 15 d. The level of IL-10 increased in all groups throughout the experiment; the FS+AG group had the lowest IL-13 concentration at 15 and 30 d. The concentration of HSP 70 increased in AG ewes at the end of the experiment and decreased in FS ewes, whereas that of HSP 90 increased in FS ewes compared with FS+AG ewes. Flaxseed supplementation was found to influence in vivo HPA activation in heat-stressed sheep, resulting in increased cortisol concentrations, probably to meet increased energy demand for thermoregulation. Flaxseed supplementation also supported Th1 response via a complex cross-talk between IL-10, IL-12, and IFN-γ production.