Merck
  • OGA Inhibition Alters Energetics and Nutrient Sensing in Alzheimer's Disease Cytoplasmic Hybrids.
OGA Inhibition Alters Energetics and Nutrient Sensing in Alzheimer's Disease Cytoplasmic Hybrids.
Journal of Alzheimer's disease : JAD (2020-12-09)
Jarrod Flax, Heather M Wilkins, Reegan Miller, Sarah Griffith, Gentry K Cork, Amy Qiang, Jeffrey Thompson, Russell H Swerdlow, Chad Slawson
ABSTRACT
Alzheimer's disease (AD) features reductions in key bioenergetic fluxes and perturbed mitochondrial function. Cytoplasmic hybrids (cybrids) generated through the transfer of AD subject mitochondria to mtDNA-depleted SH-SY5Y neuroblastoma cells recapitulate some of these features in an in vitro setting. For this study, we used the AD cybrid model to assess the impact of a nutrient-excess like-state via increasing O-GlcNAcylation on whole cell and mitochondrial homeostasis. We induced increased O-GlcNAc by treating AD and control cybrid cell lines with Thiamet G (TMG), an inhibitor of the O-GlcNAcase enzyme that mediates removal of the nutrient-dependent O-GlcNAc modification. Relative to control cybrid cell lines, AD cybrid lines showed a blunted response to TMG-induced O-GlcNAcylation. At baseline, AD cybrid cell line mitochondria showed partial activation of several proteins that help maintain bioenergetic homeostasis such as AMP-Regulated Kinase suggesting that AD mitochondria initiate a state of nutrient stress promoting energetic compensation; however, this compensation reduces the capacity of cells to respond to additional nutrient-related stresses such as TMG treatment. Also, TMG caused disruptions in acetylation and Sirtuin 3 expression, while lowing total energetic output of the cell. Together, these findings suggest that modulation of O-GlcNAc is essential for proper energetic function of the mitochondria, and AD mitochondrial capacity to handle nutrient-excess is limited.