Merck
  • Protein kinase C inhibitor anchored BRD4 PROTAC PEGylated nanoliposomes for the treatment of vemurafenib-resistant melanoma.
Protein kinase C inhibitor anchored BRD4 PROTAC PEGylated nanoliposomes for the treatment of vemurafenib-resistant melanoma.
Experimental cell research (2020-09-09)
Yige Fu, Drishti Rathod, Ketan Patel
ABSTRACT
Limited treatment options and development of resistance to targeted therapy within few months pose significant challenges in the treatment of BRAF-mutated malignant melanoma. Moreover, extensive angiogenesis and vasculogenic mimicry promote the rapid progression of disease. The purpose of this study was to develop a protein kinase C inhibitor anchored BRD4 PROTAC (ARV) loaded PEGylated nanoliposomes (LARPC). Palmitoyl-dl-carnitine chloride (PC) was used as a protein kinase C inhibitor to provide a cationic surface charge to LARPC. The formulation was characterized for particle size, zeta potential, drug release and various cell culture assays using HUVEC and vemurafenib resistant melanoma cells. The particle size of LARPC was found to be 105.25 ± 2.76 nm with a zeta potential of +26.6 ± 6.25 mV. Inhibition of angiogenesis was demonstrated by ARV and LARPC using human umbilical vein endothelial cells (HUVEC)-based matrigel basement membrane model. Additionally, LARPC demonstrated very low IC50 with promising inhibition of vasculogenic mimicry channel formation, cell migration as well as colony formation in vemurafenib-resistant melanoma cell lines. Hence, the outcome of this combination therapy indicated the suitability of LARPC as a potential and novel approach for eradicating vemurafenib-resistant melanoma.
Sigma-Aldrich
Palmitoyl-DL-carnitine chloride, powder
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder