Merck
  • Nanoscale metal-organic frameworks for x-ray activated in situ cancer vaccination.
Nanoscale metal-organic frameworks for x-ray activated in situ cancer vaccination.
Science advances (2020-10-04)
Kaiyuan Ni, Guangxu Lan, Nining Guo, August Culbert, Taokun Luo, Tong Wu, Ralph R Weichselbaum, Wenbin Lin
ABSTRACT
Cancer vaccines have been actively pursued to bolster antitumor immunity. Here, we designed nanoscale metal-organic frameworks (nMOFs) as locally activable immunotherapeutics to release danger-associated molecular patterns (DAMPs) and tumor antigens and deliver pathogen-associated molecular patterns (PAMPs) for in situ personalized cancer vaccination. When activated by x-rays, nMOFs effectively generate reactive oxygen species to release DAMPs and tumor antigens while delivering CpG oligodeoxynucleotides as PAMPs to facilitate the maturation of antigen-presenting cells. Together, DAMPs, tumor antigens, and PAMPs expand cytotoxic T cells in tumor-draining lymph nodes to reinvigorate the adaptive immune system for local tumor regression. When treated in combination with an immune checkpoint inhibitor, the local therapeutic effects of nMOF-based vaccines were extended to distant tumors via attenuating T cell exhaustion. Our work demonstrates the potential of nMOFs as x-ray-activable in situ cancer vaccines to awaken the host's innate and adaptive immune systems for systemic antitumor immunity.