Merck
CDK4/6 regulate lysosome biogenesis through TFEB/TFE3.
The Journal of cell biology (2020-07-15)
Qiuyuan Yin, Youli Jian, Meng Xu, Xiahe Huang, Niya Wang, Zhifang Liu, Qian Li, Jinglin Li, Hejiang Zhou, Lin Xu, Yingchun Wang, Chonglin Yang
ABSTRACT
Lysosomes are degradation and signaling organelles that adapt their biogenesis to meet many different cellular demands; however, it is unknown how lysosomes change their numbers for cell division. Here, we report that the cyclin-dependent kinases CDK4/6 regulate lysosome biogenesis during the cell cycle. Chemical or genetic inactivation of CDK4/6 increases lysosomal numbers by activating the lysosome and autophagy transcription factors TFEB and TFE3. CDK4/6 interact with and phosphorylate TFEB/TFE3 in the nucleus, thereby inactivating them by promoting their shuttling to the cytoplasm. During the cell cycle, lysosome numbers increase in S and G2/M phases when cyclin D turnover diminishes CDK4/6 activity. These findings not only uncover the molecular events that direct the nuclear export of TFEB/TFE3, but also suggest a mechanism that controls lysosome biogenesis in the cell cycle. CDK4/6 inhibitors promote autophagy and lysosome-dependent degradation, which has important implications for the therapy of cancer and lysosome-related disorders.