Merck
  • Cross-linkage urease nanoparticles: a high-efficiency signal-generation tag for portable pH meter-based electrochemical immunoassay of lipocalin-2 protein diagnostics.
Cross-linkage urease nanoparticles: a high-efficiency signal-generation tag for portable pH meter-based electrochemical immunoassay of lipocalin-2 protein diagnostics.
Mikrochimica acta (2020-08-07)
Ai-Li Sun, Qing-An Qi, Li-Juan Zhi
ABSTRACT
An innovative signal-transduction tag based on cross-linked urease nanoparticles (CLENP) was designed for the development of a pH meter-based immunoassay of lipocalin-2 (LCN2). The CLENP was synthesized with a typical desolvation method using ethanol as desolvation agent, followed by functionalization with polyaspartic acid. The carboxylated CLENP were used as the signal-generation tags for the labelling of secondary antibodies via the carbodiimide coupling. Upon target LCN2 introduction, a sandwich-type immune reaction was performed between capture antibody-coated plate and the labeled secondary antibody on the CLENP. The conjugated CLENP in the microplate hydrolyzed urea into ammonia (NH4+) and carbonate (CO32-), resulting in the pH change of solution, which was determined with a handheld pH meter. The pH variation was proportional to target concentration in the sample. By monitoring the pH variation of the urea solution, the level of LCN2 at a concentration as low as 5.2 pg mL-1 was evaluated. The pH meter-based electrochemical immunoassay can be utilized for mass production of miniaturized lab-on-a-chip devices with handheld pH meter, thereby opening new opportunities for protein diagnostics and biosecurity. Graphical abstract An innovative signal-transduction tag based on cross-linked urease nanoparticles was designed for high-efficiency immunoassay of lipocalin-2 with pH meter readout.
Sigma-Aldrich
Human Lipocalin-2 / NGAL ELISA Kit, for serum, plasma, cell culture supernatant and urine