Merck
  • Effectiveness in the inhibition of dapagliflozin and canagliflozin on M-type K+ current and α-methylglucoside-induced current in pituitary tumor (GH3) and pheochromocytoma PC12 cells.
Effectiveness in the inhibition of dapagliflozin and canagliflozin on M-type K+ current and α-methylglucoside-induced current in pituitary tumor (GH3) and pheochromocytoma PC12 cells.
European journal of pharmacology (2020-05-01)
Edmund Cheung So, Ping-Yen Liu, Sheng-Nan Wu
ABSTRACT
Dapagliflozin (DAPA) or canagliflozin (CANA), Na+-dependent glucose co-transporter type 2 (SGLT2) inhibitors, were used for treatment of type II diabetes mellitus. Addition of DAPA or CANA suppressed M-type K+ current (IK(M)) in pituitary tumor (GH3) and pheochromocytoma PC12 cells. The IC50 value for DAPA- or CANA-mediated inhibition of IK(M) in GH3 cells was 0.11 or 0.42 μM, respectively. The presence of DAPA (0.1 μM) shifted the steady-state activation of IK(M) to less depolarized potential without changing the gating charge of the current. During high-frequency depolarizing pulses, IK(M) magnitude was reduced by DAPA; however, DAPA-induced block of IK(M) remained effective. The amplitude of neither erg-mediated K+ current nor hyperpolarization-activated cation current in GH3 cells was modified in the presence of 1 μM DAPA. Alternatively, addition of DAPA, CANA, phlorizin or chlorotoxin effectively suppressed α-methylglucoside-(αMG-) induced current (IαMG) in GH3 cells, albeit inability of tefluthrin (activator of INa) to suppress this current. DAPA shifted the charge-voltage relation of presteady-state IαMG in a rightward and downward direction with no change in the gating charge of the IαMG. Under current-clamp recordings, subsequent additions of DAPA, but still in the continued presence of αMG, increased the firing rate of spontaneous action potentials stimulated by αMG. Our results suggested that activity of SGLT was expressed functionally in GH3 and PC12 cells. Therefore, inhibitory actions of DAPA or CANA on the amplitude and gating of IK(M) might provide a yet unidentified mechanism through which the SGLT1 or SGLT2 activity were attenuated in unclamped cells occurring in vivo.
Product Description
Supelco
Tefluthrin, PESTANAL®, analytical standard
Sigma-Aldrich
Linopirdine, ≥98% (HPLC)