Merck
  • Chemo-enzymatic synthesis, characterization, in vitro antioxidant capacity and oxidative stability studies of novel phosphatidylcholines with ω-3/ω-6 PUFAs and phenolic acids.
Chemo-enzymatic synthesis, characterization, in vitro antioxidant capacity and oxidative stability studies of novel phosphatidylcholines with ω-3/ω-6 PUFAs and phenolic acids.
Food research international (Ottawa, Ont.) (2020-04-06)
Balakrishna Marrapu, Jingjing Ma, Zhiming Geng, Swapna Nalla, Fang Liu, Pengpeng Li, Daoying Wang, Muhan Zhang, Weimin Xu
ABSTRACT
Novel phosphatidylcholines containing PUFAs and phenolic acids were synthesized from egg phosphatidylcholine (PC), PUFAs (docosahexaenoic, arachidonic and linoleic acids) and phenolic acids (caffeic, ferulic and p-coumaric acids) as substrates. The structures of modified PCs were confirmed by spectral analysis and were evaluated for antioxidant activities. The modified PCs containing caffeic and ferulic acids exhibited excellent antioxidant activities compared with butylated hydroxytoluene (BHT) and α-tocopherol. The synthesized compounds were also evaluated for the oxidative stabilities in liposome and organic solvent. The modified PCs showed more oxidative stable compared with standard PUFA-PCs and PUFA-PCs + BHT. Results showed that the oxidative stability decreased with increasing degree of unsaturation in organic solvent whereas in liposomes, increased with increasing degree of unsaturation due to tight packed configuration. In this study, phenolic acids were found to render protections for PUFAs in modified PCs from oxidation. Modified PCs may have great potential for applications in food, cosmetic and pharmaceutical industries.