Merck
  • MNK1 signaling induces an ANGPTL4-mediated gene signature to drive melanoma progression.
MNK1 signaling induces an ANGPTL4-mediated gene signature to drive melanoma progression.
Oncogene (2020-03-07)
William Yang, Elie Khoury, Qianyu Guo, Sathyen A Prabhu, Audrey Emond, Fan Huang, Christophe Gonçalves, Yao Zhan, Dany Plourde, Jessica N Nichol, Michael S Dahabieh, Wilson H Miller, Sonia Victoria Del Rincón
ABSTRACT
The BRAFV600E mutation occurs in more than 50% of cutaneous melanomas, and results in the constitutive activation of the mitogen-activated protein kinases (MAPK) pathway. MAP kinase-interacting serine/threonine-protein kinase 1 and 2 (MNK1/2) are downstream effectors of the activated MAPK pathway, and important molecular targets in invasive and metastatic cancer. Despite the well-known role of MNK1 in regulating mRNA translation, little is known concerning the impact of its aberrant activation on gene transcription. Here, we show that changes in the activity, or abundance, of MNK1 result in changes in the expression of pro-oncogenic and pro-invasive genes. Among the MNK1-upregulated genes, we identify Angiopoietin-like 4 (ANGPTL4), which in turn promotes an invasive phenotype via its ability to induce the expression of matrix metalloproteinases (MMPs). Using a pharmacologic inhibitor of MNK1/2, SEL201, we demonstrate that BRAFV600E-mutated cutaneous melanoma cells are reliant on MNK1/2 for invasion and lung metastasis.
Sigma-Aldrich
MISSION® esiRNA, targeting human ANGPTL4