Merck
  • Clues and challenges in the diagnosis of intermittent maple syrup urine disease.
Clues and challenges in the diagnosis of intermittent maple syrup urine disease.
European journal of medical genetics (2020-03-11)
Naomi Pode-Shakked, Stanley H Korman, Ben Pode-Shakked, Yuval Landau, Katya Kneller, Smadar Abraham, Avraham Shaag, Igor Ulanovsky, Suha Daas, Talya Saraf-Levy, Haike Reznik-Wolf, Asaf Vivante, Elon Pras, Shlomo Almashanu, Yair Anikster
ABSTRACT
Maple syrup urine disease is a rare autosomal-recessive aminoacidopathy, caused by deficient branched-chain 2-keto acid dehydrogenase (BCKD), with subsequent accumulation of branched-chain amino acids (BCAAs): leucine, isoleucine and valine. While most cases of MSUD are classic, some 20% of cases are non-classic variants, designated as intermediate- or intermittent-types. Patients with the latter form usually develop normally and are cognitively intact, with normal BCAA levels when asymptomatic. However, intercurrent febrile illness and catabolism may cause metabolic derailment with life-threatening neurological sequelae. Thus, early detection and dietary intervention are warranted in intermittent MSUD. We describe eight patients from four unrelated families, diagnosed with intermittent MSUD. Their presenting symptoms during metabolic crises varied from confusion and decreased consciousness, to ataxia, and acute psychosis. Molecular confirmation of MSUD was pursued via sequencing of the BCKDHA, BCKDHB and DBT genes. All affected individuals were found to harbor bi-allelic pathogenic variants in either BCKDHB or DBT. Of the seven variants, four variants in BCKDHB (p.G101D, p. V103A, p. A221D, p. Y195C) and one variant in DBT (p.K427E) were not previously described. While newborn screening programs allow for early detection of classic MSUD, cases of the intermittent form might go undetected, and present later in childhood following metabolic derailment, with an array of non-specific symptoms. Our experience with the families reported herein adds to the current knowledge regarding the phenotype and mutational spectrum of this unique inborn error of branched-chain amino acid metabolism, and underscore the high index of suspicion required for its diagnosis.