Merck
  • Production of tagatose and talose through isomerization of galactose in a buffer solution under subcritical water conditions.
Production of tagatose and talose through isomerization of galactose in a buffer solution under subcritical water conditions.
Carbohydrate research (2020-05-27)
Yuichiro Onishi, Yuya Furushiro, Yusuke Hirayama, Shuji Adachi, Takashi Kobayashi
ABSTRACT
Galactose was isomerized in pure water or in 10 mmol/L sodium phosphate buffer at 160 °C under pressurized conditions. The isomerization of galactose to tagatose and talose in phosphate buffer resulted in 14% and 1.4% yields, respectively, which were significantly higher than those obtained in subcritical pure water (0.6% and <0.1%, respectively). The effect of the temperature on isomerization was examined between 100 and 160 °C. The most remarkable isomerization was observed at 120 °C or higher. The effect of the buffer solution type was also examined. The pH drop of the treated solution was lesser in MOPS and PIPES buffers than in the phosphate buffer; however, the isomerization was less likely to occur in MOPS and PIPES buffers. The relationship between the pH drop during the reaction in phosphate buffer and the yields of tagatose and talose revealed that the isomerization proceeded only when the pH was >6.3. Our results indicate that the galactose isomerization by Lobry de Bruyn-Alberda-van Ekenstein transformation rarely occurred at low pH due to the formation of organic acids.
Sigma-Aldrich
D-(+)-Talose, ≥99.0% (HPLC)