Merck
  • Effects of neuropeptide Y on the microvasculature of human skeletal muscle.
Effects of neuropeptide Y on the microvasculature of human skeletal muscle.
Surgery (2020-06-05)
Benjamin Mirman, Ian Ikeda, Zhiqi Zhang, Yuhong Liu, Lucy Yu, Afshin Ehsan, Jun Feng, Frank Sellke
ABSTRACT
Neuropeptide Y acts directly on the vasculature as a cotransmitter with norepinephrine for an augmented contraction. Little, however, is known about the effects of neuropeptide Y on the microvasculature of human skeletal muscle. Neuropeptide Y signaling has not been studied in the setting of cardiac surgery and cardiopulmonary bypass. We investigated the role of neuropeptide Y signaling on vasomotor tone in the microvessels of human skeletal muscle, as well as the effect of cardiopulmonary bypass on neuropeptide Y-induced responsiveness. Specimens taken from intercostal muscles were collected from patients, pre- and post-cardiopulmonary bypass, undergoing coronary artery bypass grafting or cardiac valve surgery (n = 8/group). Microvessels (157 ± 47 microns) were isolated in vitro in a no-flow state. Arterial microvascular responses to a neuropeptide Y agonist, a Y1 receptor antagonist, phenylephrine, and the coadministration of neuropeptide Y and phenylephrine were examined. The abundance and localization of the Y1 receptor were measured using Western blot and immunofluorescence, respectively. Arterial microvessels showed responsiveness to the neuropeptide Y agonist (10-9 to 4 × 10-7 mol/L) both before and after cardiopulmonary bypass, reaching a 12.5% vasoconstriction from the baseline luminal diameter. With administration of the Y1 receptor antagonist after neuropeptide Y, the contractile response was eliminated (n = 3/group, P = .04). No difference in vasoconstriction was observed between pre- and post-cardiopulmonary bypass groups (P = .73). The coadministration of neuropeptide Y and phenylephrine (10-9 to 10-4 mol/L) elicited no difference in vasoconstriction (n = 7/group, P = .06 both pre- and post-cardiopulmonary bypass) when compared with phenylephrine alone (10-9 to 10-4 mol/L). No change in the protein expression or localization of the Y1 receptor was detected by Western blotting (n = 6/group, P = .44) or immunofluorescence (n = 6/group, P = .13). Neuropeptide Y induced vasoconstriction, suggesting that neuropeptide Y may play an important role in the regulation of the peripheral microvasculature. There was no change in microvascular responsiveness to neuropeptide Y after cardiopulmonary bypass nor were there any synergistic effects of neuropeptide Y on phenylephrine-induced vasoconstriction in the skeletal muscle microvasculature.