Merck
  • The influence of selective serotonin reuptake inhibitors on the plasma and brain pharmacokinetics of the simplest phenothiazine neuroleptic promazine in the rat.
The influence of selective serotonin reuptake inhibitors on the plasma and brain pharmacokinetics of the simplest phenothiazine neuroleptic promazine in the rat.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology (1999-07-28)
W A Daniel, M Syrek, J Wójcikowski
ABSTRACT
The aim of the present study was to investigate a possible impact of the three selective serotonin reuptake inhibitors (SSRIs) fluoxetine, fluvoxamine and sertraline on the pharmacokinetics of promazine in a steady state in rats. Promazine was administered twice a day for 2 weeks, alone or jointly with one of the antidepressants. Concentrations of promazine and its two main metabolites (N-desmethylpromazine and sulfoxide) in the plasma and brain were measured at 30 min and 6 and 12 h after the last dose of the drugs. All the investigated SSRIs increased the plasma and brain concentrations of promazine up to 300% of the control value, their effect being most pronounced after 30 min and 6 h. Moreover, simultaneous increases in the promazine metabolites' concentrations and in the promazine-metabolite concentration ratios were observed. In vitro studies with liver microsomes of rats treated chronically with promazine, SSRIs or their combination did not show any significant changes in the concentrations of cytochromes P-450 and b-5. However, treatment with fluoxetine, alone or in a combination with promazine, decreased the rates of promazine N-demethylation and sulfoxidation. A similar effect was observed in the case of promazine and fluvoxamine combination. Kinetic studies into promazine metabolism, carried out on control liver microsomes in the absence or presence of SSRIs added in vitro, demonstrated competitive inhibition of both N-demethylation and sulfoxidation by the antidepressants. The results of in vivo and in vitro studies indicate the following mechanisms of the observed interactions: (a) competition for an active site of promazine N-demethylase and sulfoxidase; (b) adaptive changes in cytochrome P-450, produced by chronic treatment with fluoxetine or fluvoxamine; (c) additionally, increases in the sum of concentrations of promazine+ metabolites, produced by fluoxetine and sertraline in vivo, suggest simultaneous inhibition of another, not investigated by us, metabolic pathway of promazine, e.g. hydroxylation. In conclusion, all the three SSRIs administered chronically in pharmacological doses, increase the concentrations of promazine in the blood plasma and brain of rats by inhibiting different metabolic pathways of the neuroleptic. Assuming that similar interactions occur in humans, reduced doses of phenotiazines should be considered when one of the above antidepressants is to be given jointly.