Merck
  • Electrical monitoring of photoisomerization of block copolymers intercalated into graphene sheets.
Electrical monitoring of photoisomerization of block copolymers intercalated into graphene sheets.
Nature communications (2020-03-14)
Semin Kim, Thanh-Hai Le, Yunseok Choi, Haney Lee, Eunseo Heo, Unhan Lee, Saerona Kim, Subin Chae, Yoong Ahm Kim, Hyeonseok Yoon
ABSTRACT
Insulating polymers have received little attention in electronic applications. Here, we synthesize a photoresponsive, amphiphilic block copolymer (PEO-b-PVBO) and further control the chain growth of the block segment (PVBO) to obtain different degrees of polymerization (DPs). The benzylidene oxazolone moiety in PEO-b-PVBO facilitated chain-conformational changes due to photoisomerization under visible/ultraviolet (UV) light illumination. Intercalation of the photoresponsive but electrically insulating PEO-b-PVBO into graphene sheets enabled electrical monitoring of the conformational change of the block copolymer at the molecular level. The current change at the microampere level was proportional to the DP of PVBO, demonstrating that the PEO-b-PVBO-intercalated graphene nanohybrid (PGNH) can be used in UV sensors. Additionally, discrete signals at the nanoampere level were separated from the first derivative of the time-dependent current using the fast Fourier transform (FFT). Analysis of the harmonic frequencies using the FFT revealed that the PGNH afforded sawtooth-type current flow mediated by Coulomb blockade oscillation.
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%
Sigma-Aldrich
Potassium persulfate, AR, ≥99%