Merck
  • Multiplexed derivatization strategy-based dummy molecularly imprinted polymers as sorbents for magnetic dispersive solid phase extraction of globotriaosylsphingosine prior to UHPLC-MS/MS quantitation.
Multiplexed derivatization strategy-based dummy molecularly imprinted polymers as sorbents for magnetic dispersive solid phase extraction of globotriaosylsphingosine prior to UHPLC-MS/MS quantitation.
Mikrochimica acta (2020-06-07)
Jingwen Hu, Shuyun Zhu, Shi-En Chen, Ruisheng Liu, Jing Sun, Xian-En Zhao, Huwei Liu
ABSTRACT
A new series of 9-plex chemical isotope-labeling reagents, levofloxacin-based mass tags (LMTs) named as LMT359, 360, 361, 362, 363, 373, 375, 376, and 378, was firstly designed and synthesized for the high-throughput labeling of globotriaosylsphingosine (lyso-Gb3), a disease biomarker of Fabry disease. Creatively based on derivatization strategy-dummy template technique, dummy magnetic molecularly imprinted polymers (DMMIPs) were designed and prepared using LMT387-labeled lyso-Gb3 as a dummy template. The novel DMMIP material was used as sorbents for magnetic dispersive solid-phase extraction of 9-plexed LMT derivatives of lyso-Gb3 from equally mixed derivatization solutions. The enriched 8-plexed lyso-Gb3 derivatives from 8 real samples were quantified by ultra-high-performance liquid chromatography tandem mass spectrometry in a single run using simultaneously extracted LMT359-labeled standard lyso-Gb3 as internal standards. DMMIPs were characterized by using the transmission electron microscope (TEM), Fourier transform infrared, X-ray photoelectron spectroscopy, and some other characterization techniques. TEM micrograph showed that the prepared DMMIPs had an apparent imprinting layer. Triple-recognition abilities of DMMIPs towards LMT-lyso-Gb3 mainly rely on the hydrogen bonding, electrostatic attraction, hydrophobic interaction, and boronate affinity. The imprinting factor of DMMIPs towards LMT-lyso-Gb3 was 5.1. This method shows the advantages of high selectivity (triple recognition), high sensitivity, high accuracy (recovery 93.5-108.8%), and high throughput (8 samples in a single run). The proposed method was successfully applied to the determination of lyso-Gb3 in plasma samples with spiked recoveries in the range of 95.0-102.4%. This indicates that the method is promising in bioanalysis and medical testing of lyso-Gb3 in the future. Graphical abstract Synthesis of multiplexed derivatization reagents and its correlative molecularly imprinted polymers for magnetic extraction of globotriaosylsphingosine.
Sigma-Aldrich
Glucosylsphingosine, ≥98.0% (TLC)