- Phosphoethanolamine induces caspase-independent cell death by reducing the expression of C-RAF and inhibits tumor growth in human melanoma model.
Phosphoethanolamine induces caspase-independent cell death by reducing the expression of C-RAF and inhibits tumor growth in human melanoma model.
Phosphoethanolamine (PEA) is a fundamental precursor during the biosynthesis of cell membranes phospholipids. In the past few years, it has been described as a potential antitumor agent. In previous studies, we demonstrated that PEA showed antitumor properties in vitro and in vivo in a wide range of tumor cell lines. Herein, we showed that PEA possesses cytotoxic properties and notably revealed to induce caspase-independent cell death. Of interest, we provided evidence that PEA inhibits melanoma cells proliferation through the reduction of C-RAF. Molecular docking of PEA evidenced that this compound indeed fits satisfactory in the binding site located between the dimers of C-RAF protein with 107,01 Å and score of -29,62. Also, PEA arrested A2058 cells at G2/M phase in the cell cycle. Moreover, cell proliferation, migration and adhesion capacities of A2058 cells were also inhibited by PEA. Most importantly, PEA inhibited tumor growth of melanoma tumors and prolonged survival rate of mice. Also, PEA induced a significant immune response in a syngeneic metastatic melanoma model. Taken together, these data indicate that PEA is a promising candidate for future developments in cancer field.