Merck
  • Calcitriol Prevents RAD51 Loss and cGAS-STING-IFN Response Triggered by Progerin.
Calcitriol Prevents RAD51 Loss and cGAS-STING-IFN Response Triggered by Progerin.
Proteomics (2019-12-14)
Nuria Coll-Bonfill, Rafael Cancado de Faria, Sweta Bhoopatiraju, Susana Gonzalo
ABSTRACT
Hutchinson Gilford progeria syndrome (HGPS) is a devastating accelerated aging disease caused by LMNA gene mutation. The truncated lamin A protein produced "progerin" has a dominant toxic effect in cells, causing disruption of nuclear architecture and chromatin structure, genomic instability, gene expression changes, oxidative stress, and premature senescence. It was previously shown that progerin-induced genomic instability involves replication stress (RS), characterized by replication fork stalling and nuclease-mediated degradation of stalled forks. RS is accompanied by activation of cGAS/STING cytosolic DNA sensing pathway and STAT1-regulated interferon (IFN)-like response. It is also found that calcitriol, the active hormonal form of vitamin D, rescues RS and represses the cGAS/STING/IFN cascade. Here, the mechanisms underlying RS in progerin-expressing cells and the rescue by calcitriol are explored. It is found that progerin elicits a marked downregulation of RAD51, concomitant with increased levels of phosphorylated-RPA, a marker of RS. Interestingly, calcitriol prevents RS and activation of the cGAS/STING/IFN response in part through maintenance of RAD51 levels in progerin-expressing cells. Thus, loss of RAD51 is one of the consequences of progerin expression that can contribute to RS and activation of the IFN response. Stabilization of RAD51 helps explain the beneficial effects of calcitriol in these processes.