Merck
  • Protein expression of PKCZ (Protein Kinase C Zeta), Munc18c, and Syntaxin-4 in the insulin pathway in endometria of patients with polycystic ovary syndrome (PCOS).
Protein expression of PKCZ (Protein Kinase C Zeta), Munc18c, and Syntaxin-4 in the insulin pathway in endometria of patients with polycystic ovary syndrome (PCOS).
Reproductive biology and endocrinology : RB&E (2012-03-07)
Rodrigo Rivero, Claire-Alix Garin, Paulina Ormazabal, Andrea Silva, Rodrigo Carvajal, Fernando Gabler, Carmen Romero, Margarita Vega
ABSTRACT
Polycystic Ovary Syndrome (PCOS) is an endocrine-metabolic disorder commonly associated with insulin resistance (IR). Previous studies indicate about the expression of molecules involved in the insulin pathway in endometria of women with PCOS-IR. Therefore, the aim of the present study was to evaluate the effect of insulin and testosterone in the expression of these proteins in the endometria and immortal endometrial stromal cell line (T-HESCs). We examined the protein levels of Munc18c, PKC zeta, phospho-PKC Zeta, and Syntaxin-4. Protein levels were assessed by Western Blot and/or immunohistochemistry in proliferative endometria (NPE = 6) and in PCOS endometria with insulin resistance (PCOSE-IR = 6). We also evaluated whether high concentrations of insulin (100 nM) and/or testosterone (100 nM), during a 24 h stimulatory period, affected the expression of these proteins in an immortal endometrial stromal cell line (T-HESCs). Once stimulated, proteins were extracted from cells and were assessed by Western Blot analysis. Immunocytochemistry was performed to detect AR in T-HESC cells. Western Blot data showed decreased expression (p < 0,05) of Munc18c and phospho-PKC Zeta in PCOS-IR endometria (PCOSE-IR) with respect to the control (NPE). In the in vitro study, Western Blot analysis showed decreased levels of Munc18c, PKC Zeta and phospho-PKC Zeta with the different hormonal treatments when compared to the control condition (no hormonal stimulation) (p < 0,05). The AR was present in the endometrial stromal cell line (T-HESC). The conditions of hyperinsulinism and hyperandrogenism present in PCOS-IR patients modulate the expression and/or phosphorylation of the proteins involved in the insulin pathway at the endometrial level. These data extend to the T-HESCs cells results, where insulin and testosterone exert an effect on both the expression and phosphorylation of proteins present in the pathway.