Merck
  • Heterologous overexpression of a novel halohydrin dehalogenase from Pseudomonas pohangensis and modification of its enantioselectivity by semi-rational protein engineering.
Heterologous overexpression of a novel halohydrin dehalogenase from Pseudomonas pohangensis and modification of its enantioselectivity by semi-rational protein engineering.
International journal of biological macromolecules (2020-01-01)
Feng Xue, Xiaohong Yu, Yueling Shang, Chao Peng, Lihui Zhang, Qing Xu, Aipeng Li
ABSTRACT
A gene encoding a halohydrin dehalogenase from Pseudomonas pohangensis (PpHHDH) was identified, synthesized and expressed in Escherichia coli. Subsequently, we used protein engineering to enhance the enzyme's enantioselectivity. We created two enantiocomplementary HHDH mutants, N160L and Q159L, which exhibited higher S- and R-selectivity toward PGE, respectively. The exchange of Leu at 159 for Gln led to a 2.3-fold increase in enantioselectivity (E-value of 22.2) compared to the wild-type. In addition, the N160L mutant displayed an inverted enantioselectivity (from ER = 9.8 to ES = 21.6) toward PGE. The wild-type PpHHDH and its variants were purified and characterized. They all displayed maximum activity at pH 7.5. The optimum temperature of mutant Q159L and N160L was increased from 35 °C to 40 °C. The wild-type PpHHDH and N160L mutant had good pH stability at pH 5.0-7.5, and Q159L showed an even wider range of pH tolerance, from pH 4.5 to pH 8.0. The mutants N160L and Q159L showed slightly better thermostability than wild-type PpHHDH. For most tested substrates, the two variants showed higher enantioselectivity. These findings further confirmed the importance of amino acid residues at positions 159 and 160 for the enantioselectivity of PpHHDH.
Sigma-Aldrich
Benzyl glycidyl ether, 99%