Merck
  • Synthesis of spacer-equipped phosphorylated di-, tri- and tetrasaccharide fragments of the O-specific polysaccharide of Vibrio cholerae O139.
Synthesis of spacer-equipped phosphorylated di-, tri- and tetrasaccharide fragments of the O-specific polysaccharide of Vibrio cholerae O139.
Carbohydrate research (2006-05-03)
Bart Ruttens, Pavol Kovác
ABSTRACT
The synthesis of oligosaccharide fragments of the O-specific polysaccharide of Vibrio cholerae O139 containing a 4,6-cyclic phosphate galactose residue linked to GlcNAc is described. 8-Azido-3,6-dioxaoctyl 2,3,4,6-tetra-O-acetyl-beta-D-galactopyranosyl-(1-->3)-2-acetamido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranoside, obtained by condensation of 2,3,4,6-tetra-O-acetyl-alpha-D-galactopyranosyl bromide and 8-azido-3,6-dioxaoctyl 2-acetamido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranoside, was converted to 8-azido-3,6-dioxaoctyl 3-O-benzyl-beta-D-galactopyranosyl-(1-->3)-2-acetamido-6-O-benzyl-2-deoxy-beta-D-glucopyranoside (6) by reductive opening of the acetal, followed by deacetylation and selective benzylation. Phosphorylation of 6 furnished two isomeric 4,6-cyclic 2,2,2-trichloroethyl phosphates. Glycosylation of the (S)-phosphate with 2,4-di-O-benzyl-3,6-dideoxy-alpha-L-xylo-hexopyranosyl bromide under halide-assisted conditions gave the desired tetrasaccharide, together with a trisaccharide. Global deprotection and reduction of the azide to an amine was effected by catalytic hydrogenation/hydrogenolysis to give the deprotected tetrasaccharide, which is functionalized for conjugation.
Sigma-Aldrich
Acetobromo-α-D-galactose, ≥93% (TLC)