Merck
  • Aberrant expression of the hematopoietic-restricted minor histocompatibility antigen LRH-1 on solid tumors results in efficient cytotoxic T cell-mediated lysis.
Aberrant expression of the hematopoietic-restricted minor histocompatibility antigen LRH-1 on solid tumors results in efficient cytotoxic T cell-mediated lysis.
Cancer immunology, immunotherapy : CII (2008-08-23)
Ingrid M Overes, T Henriëtte Levenga, Johanna C M Vos, Agnes van Horssen-Zoetbrood, Robbert van der Voort, Pieter H De Mulder, Theo M de Witte, Harry Dolstra
ABSTRACT
CD8(+) T cells recognizing minor histocompatibility antigens (MiHA) on solid tumor cells may mediate effective graft-versus-tumor (GVT) reactivity after allogeneic stem cell transplantation (SCT). Previously, we identified LRH-1 as a hematopoietic-restricted MiHA encoded by the P2X5 gene. Here, we report that LRH-1 is aberrantly expressed on solid tumor cells. P2X5 mRNA expression is demonstrated in a significant portion of solid tumor cell lines, including renal cell carcinoma (RCC), melanoma, colorectal carcinoma, brain cancer and breast cancer. Importantly, P2X5 gene expression was also detected in a subset of primary solid tumor specimens derived from RCC, brain cancer and breast cancer patients. Furthermore, P2X5 expressing solid tumor cells can be effectively targeted by LRH-1-specific cytotoxic T lymphocytes under inflammatory conditions. The expression of HLA-B7 and CD54 on tumor cells increases upon cytokine stimulation resulting in improved T cell activation as observed by higher levels of degranulation and enhanced tumor cell lysis. Overall, hematopoietic-restricted MiHA LRH-1 is aberrantly expressed on solid tumor cells and may be used as target in GVT-specific immunotherapy after SCT.