Merck
  • Genetic Depletion or Hyperresponsiveness of Natural Killer Cells Do Not Affect Atherosclerosis Development.
Genetic Depletion or Hyperresponsiveness of Natural Killer Cells Do Not Affect Atherosclerosis Development.
Circulation research (2017-10-20)
Wared Nour-Eldine, Jérémie Joffre, Kazem Zibara, Bruno Esposito, Andréas Giraud, Lynda Zeboudj, José Vilar, Megumi Terada, Patrick Bruneval, Eric Vivier, Hafid Ait-Oufella, Ziad Mallat, Sophie Ugolini, Alain Tedgui
ABSTRACT
Chronic inflammation is central in the development of atherosclerosis. Both innate and adaptive immunities are involved. Although several studies have evaluated the functions of natural killer (NK) cells in experimental animal models of atherosclerosis, it is not yet clear whether NK cells behave as protective or proatherogenic effectors. One of the main caveats of previous studies was the lack of specificity in targeting loss or gain of function of NK cells. We used 2 selective genetic approaches to investigate the role of NK cells in atherosclerosis: (1) Ncr1 iCre/+ R26 lsl- DTA/+ mice in which NK cells were depleted and (2) Noé mice in which NK cells are hyperresponsive. No difference in atherosclerotic lesion size was found in Ldlr-/- (low-density lipoprotein receptor null) mice transplanted with bone marrow (BM) cells from Ncr1 iCre R26R lsl- DTA , Noé, or wild-type mice. Also, no difference was observed in plaque composition in terms of collagen content, macrophage infiltration, or the immune profile, although Noé chimera had more IFN (interferon)-γ-producing NK cells, compared with wild-type mice. Then, we investigated the NK-cell selectivity of anti-asialoganglioside M1 antiserum, which was previously used to conclude the proatherogenicity of NK cells. Anti-asialoganglioside M1 treatment decreased atherosclerosis in both Ldlr-/- mice transplanted with Ncr1 iCre R26R lsl- DTA or wild-type bone marrow, indicating that its antiatherogenic effects are unrelated to NK-cell depletion, but to CD8+ T and NKT cells. Finally, to determine whether NK cells could contribute to the disease in conditions of pathological NK-cell overactivation, we treated irradiated Ldlr-/- mice reconstituted with either wild-type or Ncr1 iCre R26R lsl- DTA bone marrow with the viral mimic polyinosinic:polycytidylic acid and found a significant reduction of plaque size in NK-cell-deficient chimeric mice. Our findings, using state-of-the-art mouse models, demonstrate that NK cells have no direct effect on the natural development of hypercholesterolemia-induced atherosclerosis, but may play a role when an additional systemic NK-cell overactivation occurs.