Merck
  • Everolimus improves neuropsychiatric symptoms in a patient with tuberous sclerosis carrying a novel TSC2 mutation.
Everolimus improves neuropsychiatric symptoms in a patient with tuberous sclerosis carrying a novel TSC2 mutation.
Molecular brain (2016-05-25)
Su-Kyeong Hwang, Jae-Hyung Lee, Jung-Eun Yang, Chae-Seok Lim, Jin-A Lee, Yong-Seok Lee, Kyungmin Lee, Bong-Kiun Kaang
ABSTRACT
Tuberous sclerosis complex (TSC) is a neurocutaneous disorder characterized by multiple symptoms including neuropsychological deficits such as seizures, intellectual disability, and autism. TSC is inherited in an autosomal dominant pattern and is caused by mutations in either the TSC1 or TSC2 genes, which enhance activation of the mammalian target of rapamycin (mTOR) signaling pathway. Recent studies have suggested that mTOR inhibitors such as rapamycin can reverse TSC-associated deficits in rodent models of TSC. In addition, clinical trials are ongoing to test the efficacy of mTOR inhibitors toward the psychiatric symptoms associated with TSC. Here, we report a case study of a Korean patient with TSC, who exhibited multiple symptoms including frequent seizures, intellectual disability, language delays, and social problems. We performed whole exome sequencing and identified a novel small deletion mutation in TSC2. Expressing the novel deletion mutant in HEK293T cells significantly increased mTOR pathway activation. Furthermore, everolimus treatment showed not only reduction in SEGA size, but dramatically improved behavioral deficits including autism related behaviors in the patient. In summary, we identified a novel small deletion mutation in TSC2 associated with severe TSC in a Korean family that enhances the activation of mTOR signaling in vitro. Everolimus treatment improved behavioral deficits in the patient.