Merck
Inducing efficient cross-priming using antigen-coated yeast particles.
Journal of immunotherapy (Hagerstown, Md. : 1997) (2008-07-05)
Shanshan W Howland, Takemasa Tsuji, Sacha Gnjatic, Gerd Ritter, Lloyd J Old, Karl Dane Wittrup
ABSTRACT
Saccharomyces cerevisiae stimulates dendritic cells (DCs) and represents a promising candidate for cancer vaccine development. Effective cross-presentation of antigen delivered to DCs is necessary for successful induction of cellular immunity. Here, we present a yeast-based vaccine approach that is independent of yeast's ability to express the chosen antigen, which is instead produced separately and conjugated to the yeast cell wall. The conjugation method is site-specific (based on the SNAP-tag) and designed to facilitate antigen release in the DC phagosome and subsequent translocation for cross-presentation. We demonstrate that nonsite-specific chemical conjugation of the same protein hinders cross-presentation. Phagosomal antigen release was further expedited through the insertion of the invariant chain ectodomain as a linker, which is rapidly cleaved by Cathepsin S. The dose of delivered antigen was increased in several ways: by using yeast strains with higher surface amine densities, by using yeast hulls (cell wall fragments) instead of whole cells, and by conjugating multiple layers of antigen. The novel multilayer conjugation scheme takes advantage of Sfp phosphopantetheinyl transferase and remains site-specific; it enables the antigen dose to grow linearly with the number of layers. We show that whole yeast cells coated with 1 layer of the cancer-testis antigen NY-ESO-1 and yeast hulls bearing 3 layers were able to cross-prime naive CD8 T cells in vitro, with the latter resulting in higher frequencies of antigen-specific cells after 10 days. This cross-presentation-efficient antigen conjugation scheme is not limited to yeast and can readily be applied toward the development of other particulate vaccines.