Merck
  • Peroxynitrite decreases arrhythmias induced by ischaemia reperfusion in anaesthetized dogs, without involving mitochondrial KATP channels.
Peroxynitrite decreases arrhythmias induced by ischaemia reperfusion in anaesthetized dogs, without involving mitochondrial KATP channels.
British journal of pharmacology (2008-10-11)
A Kiss, L Juhász, I Huliák, A Végh
ABSTRACT
Exogenous peroxynitrite from nanomolar to micromolar concentrations exerts cardioprotection. Here, we have assessed its effects on ischaemia- and reperfusion-induced ventricular arrhythmias in vivo and a possible role for mitochondrial K(ATP) channels in these effects, using the channel inhibitor 5-hydroxydecanoate (5-HD). Chloralose-urethane-anaesthetized dogs were treated twice for 5 min with peroxynitrite (100 nM, by intracoronary infusions) in both the absence and presence of 5-HD (150 microg kg(-1) min(-1)), and then subjected to 25 min occlusion of the left anterior descending coronary artery. The severity of ischaemia and of arrhythmias, as well as the levels of nitrotyrosine were assessed and compared with a group of control dogs, subjected only to a 25 min occlusion and reperfusion insult. Compared with controls, infusion of peroxynitrite markedly suppressed the number of ventricular premature beats (388+/-88 vs 133+/-44), the incidence of ventricular fibrillation both during occlusion (50% vs 10%) and reperfusion (100% vs 44%), and increased survival (0% vs 50%; all P<0.05). The severity of ischaemia (epicardial ST-segment changes, inhomogeneity of electrical activation) during occlusion and nitrotyrosine levels on reperfusion were significantly less in the peroxynitrite-treated dogs than in the controls. 5-HD did not modify the cardioprotective effects of peroxynitrite. Exogenous peroxynitrite provided antiarrhythmic protection in vivo, which might have been on account of a reduction in endogenous peroxynitrite formation. This protection seemed not to be mediated through mitoK(ATP) channels.