Merck
  • Bone sialoprotein enhances migration of bone marrow stromal cells through matrices by bridging MMP-2 to alpha(v)beta3-integrin.
Bone sialoprotein enhances migration of bone marrow stromal cells through matrices by bridging MMP-2 to alpha(v)beta3-integrin.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2006-09-26)
Abdullah Karadag, Larry W Fisher
ABSTRACT
BMSCs migrate through matrix barriers and differentiate into osteoblasts. BSP enhances osteogenic cell migration through basement membrane and collagen matrices in vitro by localizing MMP-2 on the cell surface through alpha(v)beta(3)-integrin. The specific mechanisms by which bone marrow stromal cells (BMSCs) leave their primary sites, move through matrices encountered during homing to their site of final differentiation, and remove preexisting matrices in preparation for bone matrix production are not well understood. The enhanced migration of human osteoblast precursor cells through matrix barriers by bone sialoprotein (BSP) was studied by a modified Boyden-chamber assay. The bridging of normally soluble matrix metalloproteinase 2 (MMP-2) to the cell surface receptor, alpha(v)beta(3)-integrin, by BSP was analyzed by flow cytometry. BSP enhanced the in vitro passage of BMSCs and pre-osteoblasts through matrix barriers (Matrigel and denatured type I collagen) in a dose-dependent manner. An intact ArgGlyAsp (RGD) was required in the BSP for enhanced migration through the barriers but was not sufficient, as shown by the inactivity of two other SIBLING (Small Integrin-Binding LIgand, N-linked Glycoprotein) family members, osteopontin and dentin matrix protein-1. The specificity of the BSP enhancement activity was apparently caused by this molecule's ability to bridge MMP-2 to the cell surfaces. Pre-osteoblasts and their BMSC precursors may use MMP-2/BSP/integrin complexes to disrupt matrix barriers during migration to their final destinations in vivo.