Merck
  • Dietary tomato powder inhibits alcohol-induced hepatic injury by suppressing cytochrome p450 2E1 induction in rodent models.
Dietary tomato powder inhibits alcohol-induced hepatic injury by suppressing cytochrome p450 2E1 induction in rodent models.
Archives of biochemistry and biophysics (2015-01-17)
Camilla P Stice, Chun Liu, Koichi Aizawa, Andrew S Greenberg, Lynne M Ausman, Xiang-Dong Wang
ABSTRACT
Chronic and excessive alcohol consumption leads to the development of alcoholic liver disease (ALD) and greatly increases the risk of liver cancer. Induction of the cytochrome p450 2E1 (CYP2E1) enzyme by chronic and excessive alcohol intake is known to play a role in the pathogenesis of ALD. High intake of tomatoes, rich in the carotenoid lycopene, is associated with a decreased risk of chronic disease. We investigated the effects of whole tomato (tomato powder, TP), partial tomato (tomato extract, TE), and purified lycopene (LYC) against ALD development in rats. Of the three supplements, only TP reduced the severity of alcohol-induced steatosis, hepatic inflammatory foci, and CYP2E1 protein levels. TE had no effect on these outcomes and LYC greatly increased inflammatory foci in alcohol-fed rats. To further support the protective effect of TP against ALD, TP was supplemented in a carcinogen (diethylnitrosamine, DEN)-initiated alcohol-promoted mouse model. In addition to reduced steatosis and inflammatory foci, TP abolished the presence of preneoplastic foci of altered hepatocytes in DEN-injected mice fed alcohol. These reductions were associated with decreased hepatic CYP2E1 protein levels, restored levels of peroxisome proliferator-activated receptor-α and downstream gene expression, decreased inflammatory gene expression, and reduced endoplasmic reticulum stress markers. These data provide strong evidence for TP as an effective whole food prevention strategy against ALD.