Merck
  • Analysis of indoleamine 2-3 dioxygenase (IDO1) expression in breast cancer tissue by immunohistochemistry.
Analysis of indoleamine 2-3 dioxygenase (IDO1) expression in breast cancer tissue by immunohistochemistry.
Cancer immunology, immunotherapy : CII (2013-01-25)
Hatem Soliman, Bhupendra Rawal, Jimmy Fulp, Ji-Hyun Lee, Alexis Lopez, Marylin M Bui, Farah Khalil, Scott Antonia, Harris G Yfantis, Dong H Lee, Tiffany H Dorsey, Stefan Ambs
ABSTRACT
The immunosuppressive enzyme, indoleamine 2,3 dioxygenase (IDO), is overexpressed in many different tumor types including breast cancer. IDO inhibitors synergize with chemotherapy in breast cancer murine models. Characterizing IDO expression in breast cancer could define which patients receive IDO inhibitors. This study analyzed IDO protein expression in 203 breast cancer cases. The relationship between IDO, overall survival (OS), disease-specific survival (DSS), clinicopathologic, molecular, and immune tumor infiltrate factors was evaluated. Expression of IDO, estrogen receptor (ER), progesterone receptor (PR), human epithelial receptor 2, cytokeratin 5/6, epithelial growth factor receptor, phosphorylated AKT, neoangiogenesis, nitrogen oxide synthetase 2 (NOS2), cyclooxygenase 2 (COX2), FoxP3, CD8, and CD11b on archival breast cancer tissue sections was evaluated by immunohistochemistry. Associations between IDO and these markers were explored by a univariate and multivariate analysis. Survival was analyzed using Kaplan-Meier (OS) and Wilcoxon two-sample (DSS) tests. IDO expression was higher in ER+ tumors compared to ER- tumors. IDO was lower in those with higher neoangiogenesis. OS was better in ER+ patients with high IDO expression. DSS was better in node-positive patients with high IDO expression. IDO activity positively correlates with NOS2. COX2 as positively correlated with IDO on univariate but not multivariate analysis. There was a trend toward greater numbers of CD11b+ cells in IDO-low tumors. IDO protein expression is lower in ER- breast tumors with greater neoangiogenesis. Future clinical trials evaluating the synergy between IDO inhibitors and chemotherapy should take this finding into account and stratify for ER status in the trial design.