Merck
  • CHMP5 controls bone turnover rates by dampening NF-κB activity in osteoclasts.
CHMP5 controls bone turnover rates by dampening NF-κB activity in osteoclasts.
The Journal of experimental medicine (2015-07-22)
Matthew B Greenblatt, Kwang Hwan Park, Hwanhee Oh, Jung-Min Kim, Dong Yeon Shin, Jae Myun Lee, Jin Woo Lee, Anju Singh, Ki-young Lee, Dorothy Hu, Changchun Xiao, Julia F Charles, Josef M Penninger, Sutada Lotinun, Roland Baron, Sankar Ghosh, Jae-Hyuck Shim
ABSTRACT
Physiological bone remodeling requires that bone formation by osteoblasts be tightly coupled to bone resorption by osteoclasts. However, relatively little is understood about how this coupling is regulated. Here, we demonstrate that modulation of NF-κB signaling in osteoclasts via a novel activity of charged multivesicular body protein 5 (CHMP5) is a key determinant of systemic rates of bone turnover. A conditional deletion of CHMP5 in osteoclasts leads to increased bone resorption by osteoclasts coupled with exuberant bone formation by osteoblasts, resembling an early onset, polyostotic form of human Paget's disease of bone (PDB). These phenotypes are reversed by haploinsufficiency for Rank, as well as by antiresorptive treatments, including alendronate, zolendronate, and OPG-Fc. Accordingly, CHMP5-deficient osteoclasts display increased RANKL-induced NF-κB activation and osteoclast differentiation. Biochemical analysis demonstrated that CHMP5 cooperates with the PDB genetic risk factor valosin-containing protein (VCP/p97) to stabilize the inhibitor of NF-κBα (IκBα), down-regulating ubiquitination of IκBα via the deubiquitinating enzyme USP15. Thus, CHMP5 tunes NF-κB signaling downstream of RANK in osteoclasts to dampen osteoclast differentiation, osteoblast coupling and bone turnover rates, and disruption of CHMP5 activity results in a PDB-like skeletal disorder.