Merck
  • Increased intracellular Cl- concentration promotes ongoing inflammation in airway epithelium.
Increased intracellular Cl- concentration promotes ongoing inflammation in airway epithelium.
Mucosal immunology (2018-03-17)
Yi-Lin Zhang, Peng-Xiao Chen, Wei-Jie Guan, Hong-Mei Guo, Zhuo-Er Qiu, Jia-Wen Xu, Yu-Li Luo, Chong-Feng Lan, Jian-Bang Xu, Yuan Hao, Ya-Xia Tan, Ke-Nan Ye, Zhao-Rong Lun, Lei Zhao, Yun-Xin Zhu, Jiehong Huang, Wing-Hung Ko, Wei-De Zhong, Wen-Liang Zhou, Nan-Shan Zhong
ABSTRACT
Airway epithelial cells harbor the capacity of active Cl- transepithelial transport and play critical roles in modulating innate immunity. However, whether intracellular Cl- accumulation contributes to relentless airway inflammation remains largely unclear. This study showed that, in airway epithelial cells, intracellular Cl- concentration ([Cl-]i) was increased after Pseudomonas aeruginosa lipopolysaccharide (LPS) stimulation via nuclear factor-κB (NF-κB)-phosphodiesterase 4D (PDE4D)-cAMP signaling pathways. Clamping [Cl-]i at high levels or prolonged treatment with LPS augmented serum- and glucocorticoid-inducible protein kinase 1 (SGK1) phosphorylation and subsequently triggered NF-κB activation in airway epithelial cells, whereas inhibition of SGK1 abrogated airway inflammation in vitro and in vivo. Furthermore, Cl--SGK1 signaling pathway was pronouncedly activated in patients with bronchiectasis, a chronic airway inflammatory disease. Conversely, hydrogen sulfide (H2S), a sulfhydryl-containing gasotransmitter, confers anti-inflammatory effects through decreasing [Cl-]i via activation of cystic fibrosis transmembrane conductance regulator (CFTR). Our study confirms that intracellular Cl- is a crucial mediator of sustained airway inflammation. Medications that abrogate excessively increased intracellular Cl- may offer novel targets for the management of airway inflammatory diseases.