Merck
Targeting Semaphorin 3C in Prostate Cancer With Small Molecules.
Journal of the Endocrine Society (2018-12-12)
Chung C W Lee, Ravi Shashi Nayana Munuganti, James W Peacock, Kush Dalal, Ivy Z F Jiao, Ashley Shepherd, Liangliang Liu, Kevin J Tam, Colin G Sedgwick, Satyam Bhasin, Kevin C K Lee, Luke Gooding, Benjamin Vanderkruk, Tabitha Tombe, Yifan Gong, Martin E Gleave, Artem Cherkasov, Christopher J Ong
ABSTRACT
Despite the amenability of early-stage prostate cancer to surgery and radiation therapy, locally advanced and metastatic prostate cancer is clinically problematic. Chemical castration is often used as a first-line therapy for advanced disease, but progression to the castration-resistant prostate cancer phase occurs with dependable frequency, largely through mutations to the androgen receptor (AR), aberrant AR signaling, and AR-independent mechanisms, among other causes. Semaphorin 3C (SEMA3C) is a secreted signaling protein that is essential for cardiac and neuronal development and has been shown to be regulated by the AR, to drive epithelial-to-mesenchymal transition and stem features in prostate cells, to activate receptor tyrosine kinases, and to promote cancer progression. Given that SEMA3C is linked to several key aspects of prostate cancer progression, we set out to explore SEMA3C inhibition by small molecules as a prospective cancer therapy. A homology-based SEMA3C protein structure was created, and its interaction with the neuropilin (NRP)-1 receptor was modeled to guide the development of the corresponding disrupting compounds. Experimental screening of 146 in silico‒identified molecules from the National Cancer Institute library led to the discovery of four promising candidates that effectively bind to SEMA3C, inhibit its association with NRP1, and attenuate prostate cancer growth. These findings provide proof of concept for the feasibility of inhibiting SEMA3C with small molecules as a therapeutic approach for prostate cancer.