Merck
  • Cold acclimation causes fiber type-specific responses in glucose and fat metabolism in rat skeletal muscles.
Cold acclimation causes fiber type-specific responses in glucose and fat metabolism in rat skeletal muscles.
Scientific reports (2017-11-15)
Diane M Sepa-Kishi, Yass Sotoudeh-Nia, Ayesha Iqbal, George Bikopoulos, Rolando B Ceddia
ABSTRACT
This study investigated fiber type-specific metabolic responses and the molecular mechanisms that regulate glucose and fat metabolism in oxidative and glycolytic muscles upon cold acclimation. Male Wistar rats were exposed to cold (4 °C) for 7 days, and then glycogen synthesis and content, glucose and palmitate oxidation, and the molecular mechanisms underlying these metabolic pathways were assessed in soleus (Sol), extensor digitorum longus (EDL), and epitrochlearis (Epit) muscles. Cold acclimation increased glycogen synthesis, glycogen content, glucose oxidation, and reduced glycogen synthase (GS) phosphorylation only in Sol muscles. Protein kinase B (AKT), glycogen synthase kinase 3 (GSK3), and AMP-activated protein kinase (AMPK) phosphorylation increased in all three muscles upon cold acclimation. Cold acclimation increased palmitate oxidation, gene expression of the transcriptional co-activator Pgc-1α, lipoprotein lipase (Lpl), fatty acid transporter (Cd36), and Sarco/endoplasmic reticulum Ca2+-ATPase (Serca) in Sol, EDL, and Epit muscles. Sarcolipin was only detected and had its content increased in Sol muscles. In conclusion, cold-induced thermogenesis activated similar signaling pathways in oxidative and glycolytic muscles, but the metabolic fate of glucose differed in skeletal muscles with distinct fiber type composition. Furthermore, only muscles rich in type I fibers appeared to have the capacity for sarcolipin-mediated SERCA uncoupling.