Merck
  • Regulation of cell division of mature B cells by ionomycin and phorbol ester.
Regulation of cell division of mature B cells by ionomycin and phorbol ester.
Journal of immunology (Baltimore, Md. : 1950) (1992-03-15)
K M Kim, T Ishigami, D Hata, K Yamaoka, M Mayumi, H Mikawa
ABSTRACT
The growth of a human B lymphoma cell line B104, an experimental model for mature B cells, was inhibited by ionomycin but not 12-O-tetradecanoylphorbol-13-acetate (TPA). Ionomycin inhibited B104 cells from entering into the M phase of the cell cycle without affecting DNA synthesis. The inhibition of cell division of B104 cells by ionomycin occurred within 24 h after stimulation. Because such a mode of action resembles that of anti-IgM antibodies, signals transduced by Ca2+ may be responsible for the inhibition of cell division of B104 cells by anti-IgM antibodies. Indeed, EGTA suppressed the inhibition of cell division of B104 cells caused not only by ionomycin, but also by anti-IgM antibody. Although TPA itself did not have any ability to promote the growth of B104 cells, it could cancel the inhibition of cell division of B104 cells by ionomycin and increase the proportion of B104 cells entering into the M phase of the cell cycle. Staphylococcus aureus Cowan I causes the greatest proliferation of normal human peripheral blood B cells during the period from 48 to 72 h after stimulation. When ionomycin was added to S. aureus Cowan I-stimulated peripheral blood B cells at 48 h of culture, it inhibited cell division during this period without affecting DNA synthesis. In the presence of TPA, this activity of ionomycin was suppressed, and the proportion of M-phase cells increased. These results suggest that cell division of mature B cells is regulated by the signals mediated by Ca2+ and protein kinase C in a mode quite different from that of regulation of DNA synthesis.