Merck
  • Ergodicity breaking on the neuronal surface emerges from random switching between diffusive states.
Ergodicity breaking on the neuronal surface emerges from random switching between diffusive states.
Scientific reports (2017-07-16)
Aleksander Weron, Krzysztof Burnecki, Elizabeth J Akin, Laura Solé, Michał Balcerek, Michael M Tamkun, Diego Krapf
ABSTRACT
Stochastic motion on the surface of living cells is critical to promote molecular encounters that are necessary for multiple cellular processes. Often the complexity of the cell membranes leads to anomalous diffusion, which under certain conditions it is accompanied by non-ergodic dynamics. Here, we unravel two manifestations of ergodicity breaking in the dynamics of membrane proteins in the somatic surface of hippocampal neurons. Three different tagged molecules are studied on the surface of the soma: the voltage-gated potassium and sodium channels Kv1.4 and Nav1.6 and the glycoprotein CD4. In these three molecules ergodicity breaking is unveiled by the confidence interval of the mean square displacement and by the dynamical functional estimator. Ergodicity breaking is found to take place due to transient confinement effects since the molecules alternate between free diffusion and confined motion.