Merck
  • Automated adherent cell elimination by a high-speed laser mediated by a light-responsive polymer.
Automated adherent cell elimination by a high-speed laser mediated by a light-responsive polymer.
Communications biology (2018-12-12)
Yohei Hayashi, Junichi Matsumoto, Shohei Kumagai, Kana Morishita, Long Xiang, Yohei Kobori, Seiji Hori, Masami Suzuki, Toshiyuki Kanamori, Kazuhiro Hotta, Kimio Sumaru
ABSTRACT
Conventional cell handling and sorting methods require manual labor, which decreases both cell quality and quantity. To purify adherent cultured cells, cell purification technologies that are high throughput without dissociation and can be utilized in an on-demand manner are expected. Here, we developed a Laser-induced, Light-responsive-polymer-Activated, Cell Killing (LiLACK) system that enables high-speed and on-demand adherent cell sectioning and purification. This system employs a visible laser beam, which does not kill cells directly, but induces local heat production through the trans-cis-trans photo-isomerization of azobenzene moieties. Using this system in each passage for sectioning, human induced pluripotent stem cells (hiPSCs) maintained their pluripotency and self-renewal during long-term culture. Furthermore, combined with deep machine-learning analysis on fluorescent and phase contrast images, a label-free and automatic cell processing system has been developed by eliminating unwanted spontaneously differentiated cells in undifferentiated hiPSC culture conditions.