Merck
  • Inhibition of Obtusifolin on retinal pigment epithelial cell growth under hypoxia.
Inhibition of Obtusifolin on retinal pigment epithelial cell growth under hypoxia.
International journal of ophthalmology (2019-10-23)
Li-Fei Wang, Zhong-Yang Yan, Ya-Lin Li, Yan-Hui Wang, Sheng-Juan Zhang, Xin Jia, Lu Lu, Yan-Xia Shang, Xin Wang, Yun-Huan Li, Shan-Yu Li
ABSTRACT
To explore the effect of Obtusifolin on retinal pigment epithelial cell growth under hypoxia. In vitro chemical hypoxia model of ARPE-19 cells was established using cobalt chloride (CoCl2). Cell viability was tested by cell counting kit-8 (CCK-8) assay. Western blot and real-time quantitative polymerase chain reaction were applied to detect proteins and mRNAs respectively. Flow cytometry was used to examine the cell cycle. Secretion of vascular endothelial growth factor (VEGF) was tested by using enzyme linked immunosorbent assay (ELISA). Under the chemical hypoxia model established by CoCl2, hypoxia inducible factor-1α (HIF-1α) mRNA and protein levels was up-regulated. Cell viability was increased and the proportion of S phase was higher. Obtusifolin could reduce cell viability under hypoxic conditions and arrest cells in G1 phase. Obtusifolin reduced the expression of Cyclin D1 and proliferating cell nuclear antigen (PCNA) in the hypoxic environment and increased the expression of p53 and p21. The levels of VEGF, VEGFR2 and eNOS proteins and mRNA were significantly increased under hypoxia while Obtusifolin inhibited the increasing. Obtusifolin can inhibit cell growth under hypoxic conditions and down-regulate HIF-1/VEGF/eNOS secretions in ARPE-19 cells.