Merck
  • Postexercise improvement in glucose uptake occurs concomitant with greater γ3-AMPK activation and AS160 phosphorylation in rat skeletal muscle.
Postexercise improvement in glucose uptake occurs concomitant with greater γ3-AMPK activation and AS160 phosphorylation in rat skeletal muscle.
American journal of physiology. Endocrinology and metabolism (2018-08-22)
Haiyan Wang, Edward B Arias, Mark W Pataky, Laurie J Goodyear, Gregory D Cartee
ABSTRACT
A single exercise session can increase insulin-stimulated glucose uptake (GU) by skeletal muscle, concomitant with greater Akt substrate of 160 kDa (AS160) phosphorylation on Akt-phosphosites (Thr642 and Ser588) that regulate insulin-stimulated GU. Recent research using mouse skeletal muscle suggested that ex vivo 5-aminoimidazole-4-carboxamide ribonucleotide (AICAR) or electrically stimulated contractile activity-inducing increased γ3-AMPK activity and AS160 phosphorylation on a consensus AMPK-motif (Ser704) resulted in greater AS160 Thr642 phosphorylation and GU by insulin-stimulated muscle. Our primary goal was to determine whether in vivo exercise that increases insulin-stimulated GU in rat skeletal muscle would also increase γ3-AMPK activity and AS160 site-selective phosphorylation (Ser588, Thr642, and Ser704) immediately postexercise (IPEX) and/or 3 h postexercise (3hPEX). Epitrochlearis muscles isolated from sedentary and exercised (2-h swim exercise; studied IPEX and 3hPEX) rats were incubated with 2-deoxyglucose to determine GU (without insulin at IPEX; without or with insulin at 3hPEX). Muscles were also assessed for γ1-AMPK activity, γ3-AMPK activity, phosphorylated AMPK (pAMPK), and phosphorylated AS160 (pAS160). IPEX versus sedentary had greater γ3-AMPK activity, pAS160 (Ser588, Thr642, Ser704), and GU with unaltered γ1-AMPK activity. 3hPEX versus sedentary had greater γ3-AMPK activity, pAS160 Ser704, and GU with or without insulin; greater pAS160 Thr642 only with insulin; and unaltered γ1-AMPK activity. These results using an in vivo exercise protocol that increased insulin-stimulated GU in rat skeletal muscle are consistent with the hypothesis that in vivo exercise-induced enhancement of γ3-AMPK activation and AS160 Ser704 IPEX and 3hPEX are important for greater pAS160 Thr642 and enhanced insulin-stimulated GU by skeletal muscle.