Merck
  • Distinctive T cell-suppressive signals from nuclearized type 1 sphingosine 1-phosphate G protein-coupled receptors.
Distinctive T cell-suppressive signals from nuclearized type 1 sphingosine 1-phosphate G protein-coupled receptors.
The Journal of biological chemistry (2006-11-24)
Jia-Jun Liao, Mei-Chuan Huang, Markus Graler, Yong Huang, Hong Qiu, Edward J Goetzl
ABSTRACT
Sphingosine 1-phosphate (S1P) generated by cells of innate immunity and the type 1 S1P G protein-coupled receptor (S1P(1)) on mobile T cells constitute a major system for control of lymphoid organ traffic and tissue migration of T cells. Now we show that T cell activation mediated by the T cell antigen receptor translocates plasma membrane S1P(1) to nuclear envelope membranes for association there with G(i/o), Erk (1/2), and other proteins that plasma membrane S1P(1) uses to signal T cell proliferation. However, nuclear S1P(1) and plasma membrane S1P(1) transduce opposite effects of S1P on T cell proliferation and relevant signaling as exemplified by respective decreases and increases in T cell nuclear concentrations of both phospho-Erk and active (phosphorylated) c-Jun. T cell antigen receptor-mediated activation of T cells therefore both eliminates migration responses to S1P by down-regulation of plasma membrane S1P(1) and translocates the S1P-S1P(1) axis into the nuclear domain where signals are directed to transcriptional control of immune functions other than migration.