Merck
  • The Aged Microenvironment Influences the Tumorigenic Potential of Malignant Prostate Epithelial Cells.
The Aged Microenvironment Influences the Tumorigenic Potential of Malignant Prostate Epithelial Cells.
Molecular cancer research : MCR (2018-09-19)
Daniella Bianchi-Frias, Mamatha Damodarasamy, Susana A Hernandez, Rui M Gil da Costa, Funda Vakar-Lopez, Ilsa M Coleman, May J Reed, Peter S Nelson
ABSTRACT
The incidence of prostate cancer is directly linked to age, but age-associated changes that facilitate prostate cancer development and progression are poorly understood. This study investigated age-related changes in the prostate microenvironment for their influence on prostate cancer behavior. Prostate cancer cells implanted orthotopically into the prostate demonstrated accelerated tumor growth in aged compared with young mice. Metastatic lesions following intravenous injection were also more numerous in aged mice. Tumors from young and aged mice showed no significant differences concerning their proliferation index, apoptosis, or angiogenesis. However, analysis of tumor-infiltrating immune cells by IHC and RNA sequencing (RNA-seq) revealed elevated numbers of macrophages in prostates from aged mice, which are quickly polarized towards a phenotype resembling protumorigenic tumor-associated macrophages upon tumor cell engraftment. Older patients with prostate cancer (>60 years old) in The Cancer Genome Atlas Prostate Adenocarcinoma (TCGA-PRAD) dataset displayed higher expression of macrophage markers (CD163 and VSIG4) which associated with higher rates of biochemical relapse. Remodeling of the collagenous extracellular matrix (ECM) was associated with prostate cancer growth and invasion in the aged microenvironment. Moreover, the collagen matrix extracted from aged mice enhanced the invasiveness and proliferation of prostate cancer cells in vitro. Together, these results demonstrate that the aged prostatic microenvironment can regulate the growth and metastasis of malignant prostate cells, highlighting the role of resident macrophages and their polarization towards a protumorigenic phenotype, along with remodeling of the ECM. IMPLICATIONS: These findings demonstrate the importance of age-associated tumor microenvironment alterations in regulating key aspects of prostate cancer progression.