Merck
  • Negative regulation of amino acid signaling by MAPK-regulated 4F2hc/Girdin complex.
Negative regulation of amino acid signaling by MAPK-regulated 4F2hc/Girdin complex.
PLoS biology (2018-03-15)
Liang Weng, Yi-Peng Han, Atsushi Enomoto, Yasuyuki Kitaura, Shushi Nagamori, Yoshikatsu Kanai, Naoya Asai, Jian An, Maki Takagishi, Masato Asai, Shinji Mii, Takashi Masuko, Yoshiharu Shimomura, Masahide Takahashi
ABSTRACT
Amino acid signaling mediated by the activation of mechanistic target of rapamycin complex 1 (mTORC1) is fundamental to cell growth and metabolism. However, how cells negatively regulate amino acid signaling remains largely unknown. Here, we show that interaction between 4F2 heavy chain (4F2hc), a subunit of multiple amino acid transporters, and the multifunctional hub protein girders of actin filaments (Girdin) down-regulates mTORC1 activity. 4F2hc interacts with Girdin in mitogen-activated protein kinase (MAPK)- and amino acid signaling-dependent manners to translocate to the lysosome. The resultant decrease in cell surface 4F2hc leads to lowered cytoplasmic glutamine (Gln) and leucine (Leu) content, which down-regulates amino acid signaling. Consistently, Girdin depletion augments amino acid-induced mTORC1 activation and inhibits amino acid deprivation-induced autophagy. These findings uncovered the mechanism underlying negative regulation of amino acid signaling, which may play a role in tightly regulated cell growth and metabolism.