Merck
  • Childhood stress impairs social function through AVP-dependent mechanisms.
Childhood stress impairs social function through AVP-dependent mechanisms.
Translational psychiatry (2019-12-11)
Nichola M Brydges, Jessica Hall, Caroline Best, Lowenna Rule, Holly Watkin, Amanda J Drake, Catrin Lewis, Kerrie L Thomas, Jeremy Hall
ABSTRACT
Impaired social function is a core feature of many psychiatric illnesses. Adverse experiences during childhood increase risk for mental illness, however it is currently unclear whether stress early in life plays a direct role in the development of social difficulties. Using a rat model of pre-pubertal stress (PPS), we investigated effects on social behaviour, oxytocin and arginine vasopressin (AVP) in the periphery (plasma) and centrally in the paraventricular and supraoptic hypothalamic nuclei. We also explored social performance and AVP expression (plasma) in participants with borderline personality disorder (BPD) who experienced a high incidence of childhood stress. Social behaviour was impaired and AVP expression increased in animals experiencing PPS and participants with BPD. Behavioural deficits in animals were rescued through administration of the AVPR1a antagonist Relcovaptan (SR49059). AVP levels and recognition of negative emotions were significantly correlated in BPD participants only. In conclusion, early life stress plays a role in the precipitation of social dysfunction, and AVP mediates at least part of this effect.
Sigma-Aldrich
SR 49059, ≥97% (HPLC)