Merck
  • Deciphering the dual effect of lipopolysaccharides from plant pathogenic Pectobacterium.
Deciphering the dual effect of lipopolysaccharides from plant pathogenic Pectobacterium.
Plant signaling & behavior (2015-03-12)
Kettani-Halabi Mohamed, Tran Daniel, Dauphin Aurélien, Hayat El-Maarouf-Bouteau, Errakhi Rafik, Delphine Arbelet-Bonnin, Bernadette Biligui, Val Florence, Ennaji Moulay Mustapha, Bouteau François
ABSTRACT
Lipopolysaccharides (LPS) are a component of the outer cell surface of almost all Gram-negative bacteria and play an essential role for bacterial growth and survival. Lipopolysaccharides represent typical microbe-associated molecular pattern (MAMP) molecules and have been reported to induce defense-related responses, including the expression of defense genes and the suppression of the hypersensitive response in plants. However, depending on their origin and the challenged plant, LPS were shown to have complex and different roles. In this study we showed that LPS from plant pathogens Pectobacterium atrosepticum and Pectobacterium carotovorum subsp. carotovorum induce common and different responses in A. thaliana cells when compared to those induced by LPS from non-phytopathogens Escherichia coli and Pseudomonas aeruginosa. Among common responses to both types of LPS are the transcription of defense genes and their ability to limit of cell death induced by Pectobacterium carotovorum subsp carotovorum. However, the differential kinetics and amplitude in reactive oxygen species (ROS) generation seemed to regulate defense gene transcription and be determinant to induce programmed cell death in response to LPS from the plant pathogenic Pectobacterium. These data suggest that different signaling pathways could be activated by LPS in A. thaliana cells.