Merck
  • Efficient 263 GHz magic angle spinning DNP at 100 K using solid-state diode sources.
Efficient 263 GHz magic angle spinning DNP at 100 K using solid-state diode sources.
Solid state nuclear magnetic resonance (2019-04-10)
Ivan V Sergeyev, Fabien Aussenac, Armin Purea, Christian Reiter, Eric Bryerton, Steven Retzloff, Jeffrey Hesler, Leo Tometich, Melanie Rosay
ABSTRACT
The development of new, high-frequency solid-state diode sources capable of operating at 263 GHz, together with an optimized stator design for improved millimeter-wave coupling to the NMR sample, have enabled low-power DNP experiments at 263 GHz/400 MHz. With 250 mW output power, signal enhancements as high as 120 are achieved on standard samples - approximately 1/3 of the maximal enhancement available with high-power gyrotrons under similar conditions. Diode-based sources have a number of advantages over vacuum tube devices: they emit a pure mode, can be rapidly frequency-swept over a wide range of frequencies, have reproducible output power over this range, and have excellent output stability. By virtue of their small size, low thermal footprint, and lack of facility requirements, solid-state diodes are also considerably cheaper to operate and maintain than high-power vacuum tube devices. In light of these features, and anticipating further improvements in terms of available output power, solid-state diodes are likely to find widespread use in DNP and contribute to further advances in the field.